This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06931925_m1
See other BATF2 GE Assays ›
Gene Symbol:
BATF2
Gene Name:
basic leucine zipper ATF-like transcription factor 2
Gene Aliases:
-
Chromosome Location:
Chr.2: 7180019 - 7188996 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06931925_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BATF2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003122550.4 XP_003122598.1 431 104
XM_005660719.3 XP_005660776.1 445 104
XM_013994125.2 XP_013849579.1 458 104

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_003122550.4, XM_005660719.3, XM_013994125.2 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Sus scrofa

Panther Classification:

Molecular Function -

basic leucine zipper transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline