This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06931964_m1
See other MIER1 GE Assays ›
Gene Symbol:
MIER1
Gene Name:
MIER1 transcriptional regulator
Gene Aliases:
-
Chromosome Location:
Chr.6: 145609619 - 145680063 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06931964_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIER1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021096496.1 XP_020952155.1 1156 79
XM_021096497.1 XP_020952156.1 992 79
XM_021096498.1 XP_020952157.1 1085 79
XM_021096499.1 XP_020952158.1 1224 79
XM_021096500.1 XP_020952159.1 1090 79
XM_021096501.1 XP_020952160.1 1170 79
XM_021096502.1 XP_020952161.1 1238 79
XM_021096503.1 XP_020952162.1 1173 79
XM_021096505.1 XP_020952164.1 1051 79
XM_021096506.1 XP_020952165.1 1380 79
XM_021096507.1 XP_020952166.1 1196 79
XM_021096508.1 XP_020952167.1 1139 79
XM_021096509.1 XP_020952168.1 1057 79
XM_021096510.1 XP_020952169.1 1092 79
XM_021096511.1 XP_020952170.1 862 79
XM_021096512.1 XP_020952171.1 1273 79

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Sus scrofa

Panther Classification:

Molecular Function -

homeodomain transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline