This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06932309_m1
See other GPRASP1 GE Assays ›
Gene Symbol:
GPRASP1
Gene Name:
G protein-coupled receptor associated sorting protein 1
Gene Aliases:
-
Chromosome Location:
Chr.X: 83765210 - 83810090 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06932309_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPRASP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021079926.1 XP_020935585.1 470 82
XM_021079927.1 XP_020935586.1 470 82
XM_021079928.1 XP_020935587.1 624 82
XM_021079929.1 XP_020935588.1 370 82
XM_021079930.1 XP_020935589.1 361 82
XM_021079931.1 XP_020935590.1 411 82
XM_021079932.1 XP_020935591.1 515 82

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_021079926.1, XM_021079927.1, XM_021079928.1, XM_021079929.1, XM_021079930.1, XM_021079931.1, XM_021079932.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 3' Most Sus scrofa

Panther Classification:

Molecular Function -

basic helix-loop-helix transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0