This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06932413_m1
See other GCNT3 GE Assays ›
Gene Symbol:
GCNT3
Gene Name:
glucosaminyl (N-acetyl) transferase 3, mucin type
Gene Aliases:
-
Chromosome Location:
Chr.1: 112424212 - 112540984 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06932413_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GCNT3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021094501.1 XP_020950160.1 125 76

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_021094501.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Sus scrofa

Panther Classification:

Molecular Function -

glycosyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline