This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06933039_g1
See other NTHL1 GE Assays ›
Gene Symbol:
NTHL1
Gene Name:
nth like DNA glycosylase 1
Gene Aliases:
-
Chromosome Location:
Chr.3: 39935619 - 39941718 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06933039_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NTHL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003124729.4 XP_003124777.1 850 65
XM_013987820.2 XP_013843274.1 989 65
XM_013987821.2 XP_013843275.1 978 65
XM_013987822.2 XP_013843276.1 960 65
XM_013987823.2 XP_013843277.1 857 65
XM_021086780.1 XP_020942439.1 950 65
XM_021086781.1 XP_020942440.1 976 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Sus scrofa

Panther Classification:

Molecular Function -

DNA metabolism protein DNA glycosylase nucleic acid metabolism protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline