This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06933921_g1
See other TP73 GE Assays ›
Gene Symbol:
TP73
Gene Name:
tumor protein p73
Gene Aliases:
-
Chromosome Location:
Chr.6: 65169770 - 65228997 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06933921_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TP73
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021095215.1 XP_020950874.1 777 62
XM_021095216.1 XP_020950875.1 776 62
XM_021095217.1 XP_020950876.1 776 62
XM_021095218.1 XP_020950877.1 746 62
XM_021095219.1 XP_020950878.1 817 62
XM_021095220.1 XP_020950879.1 978 62
XM_021095221.1 XP_020950880.1 944 62
XM_021095222.1 XP_020950881.1 682 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Sus scrofa

Panther Classification:

Molecular Function -

P53-like transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0