This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06937654_s1
See other GCNT1 GE Assays ›
Gene Symbol:
GCNT1
Gene Name:
glucosaminyl (N-acetyl) transferase 1
Gene Aliases:
-
Chromosome Location:
Chr.1: 229429020 - 229602834 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06937654_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GCNT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003480538.4 XP_003480586.1 826 136
XM_021065059.1 XP_020920718.1 1025 136
XM_021065062.1 XP_020920721.1 1369 136
XM_021065067.1 XP_020920726.1 1043 136
XM_021065073.1 XP_020920732.1 864 136
XM_021065082.1 XP_020920741.1 800 136
GenBank mRNA AK236004.1 - 1 326 136
AK395933.1 - 3 761 136

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

glycosyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0