This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06938405_s1
See other UNK GE Assays ›
Gene Symbol:
UNK
Gene Name:
unk zinc finger
Gene Aliases:
-
Chromosome Location:
Chr.12: 5592502 - 5628249 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06938405_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UNK
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003357967.4 XP_003358015.1 2696 101
XM_005653885.3 XP_005653942.1 2743 101
XM_013979242.2 XP_013834696.1 3098 101
XM_013979243.2 XP_013834697.1 3037 101
XM_013979247.2 XP_013834701.1 2670 101
XM_013979250.2 XP_013834704.1 2707 101
XM_021066619.1 XP_020922278.1 2637 101
XM_021066621.1 XP_020922280.1 3147 101
GenBank mRNA AK348443.1 - 1 114 101

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline