This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06942363_m1
See other ATOX1 GE Assays ›
Gene Symbol:
ATOX1
Gene Name:
antioxidant 1 copper chaperone
Gene Aliases:
-
Chromosome Location:
Chr.16: 71299321 - 71314598 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06942363_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATOX1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001167641.2 NP_001161113.1 1 - 2 113 87
GenBank mRNA AK234519.1 - 1 - 2 64 87
AK399932.1 - 1 - 2 115 87

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001167641.2 transcripts.

Set Membership:

Amplicon length between 86 and 100 Sus scrofa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0