This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06942525_m1
See other TMED3 GE Assays ›
Gene Symbol:
TMED3
Gene Name:
transmembrane p24 trafficking protein 3
Gene Aliases:
-
Chromosome Location:
Chr.7: 48431629 - 48442247 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06942525_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMED3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001204363.1 NP_001191292.1 2 - 3 499 155
GenBank mRNA AK235393.1 - 2 - 3 498 155
AK239535.1 - 2 - 3 499 155
AK393064.1 - 2 - 3 536 155
AK397392.1 - 2 - 3 470 155

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

vesicle coat protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0