This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06942695_m1
See other RAC1 GE Assays ›
Gene Symbol:
RAC1
Gene Name:
Rac family small GTPase 1
Gene Aliases:
-
Chromosome Location:
Chr.3: 4781776 - 4803510 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06942695_m1
Size
Availability Inventoried
Catalog # 4331182
Price 2,694.00
Your Price
Online offer:
2694.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RAC1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001243585.1 NP_001230514.1 2 - 3 322 77
XM_021085753.1 XP_020941412.1 312 77
GenBank mRNA AK231937.1 - 2 - 3 322 77
AK391982.1 - 2 - 3 317 77
AY610273.1 - 2 - 3 111 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Sus scrofa Inventoried Probe spans exons

Panther Classification:

Molecular Function -

small GTPase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline