This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ta04094782_g1
See other EIF4E GE Assays ›
Gene Symbol:
eIF4E
Gene Name:
eukaryotic initiation factor 4E p26 subunit
Gene Aliases:
-
Chromosome Location:
-
Species:
Triticum aestivum
Species Specific ID (Flybase ID):
-
Assay ID Ta04094782_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: eIF4E
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA Z12616.2 - 349 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Triticum aestivum

Panther Classification:

Molecular Function -

translation factor translation initiation factor translational protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0