This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Vv04086565_g1
See other AP3 GE Assays ›
Gene Symbol:
AP3
Gene Name:
flowering-related B-class MADS-box protein APETALA3
Gene Aliases:
GSVIVT00014506001
Chromosome Location:
Chr.18: 11366348 - 11372377 on Build 12X
Species:
Vitis vinifera
Species Specific ID (Flybase ID):
-
Assay ID Vv04086565_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AP3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA EF418603.1 - 2 - 3 259 88
GU133632.1 - 2 - 3 259 88

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Vitis vinifera Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0