This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03682807_g1
See other PDHA1 GE Assays ›
Gene Symbol:
pdha1
Gene Name:
pyruvate dehydrogenase E1 alpha 1 subunit
Gene Aliases:
pdha, pdha1-a, pdha1-b, pdhce1a, phe1a
Chromosome Location:
Chr.2: 38914636 - 38926121 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03682807_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: pdha1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001001197.1 NP_001001197.1 5 - 6 506 80
NM_001017072.2 NP_001017072.1 4 - 5 464 80
XM_012957026.2 XP_012812480.1 5 - 6 492 80
XM_012957027.2 XP_012812481.1 5 - 6 610 80
GenBank mRNA BC067306.1 - 5 - 6 506 80
CR761539.2 - 4 - 5 464 80

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0