This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03683552_m1
See other UBA5 GE Assays ›
Gene Symbol:
uba5
Gene Name:
ubiquitin like modifier activating enzyme 5
Gene Aliases:
ube1dc1
Chromosome Location:
Chr.6: 71287431 - 71310516 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03683552_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: uba5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001004790.1 NP_001004790.1 9 - 10 1163 76
XM_018094610.1 XP_017950099.1 8 - 9 1668 76
XM_018094611.1 XP_017950100.1 8 - 9 1665 76
XM_018094612.1 XP_017950101.1 9 - 10 1750 76
GenBank mRNA BC074525.1 - 9 - 10 1163 76
CR760171.2 - 9 - 10 1071 76

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gqkn5:80/100.66.76.113:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0