This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03683886_g1
See other NECTIN2 GE Assays ›
Gene Symbol:
nectin2 LOC108645444
Gene Name:
nectin cell adhesion molecule 2
nectin-2-like
Gene Aliases:
cd111, cd112, clped1, ed4, higr, hveb, hvec, nectin-1, nectin-2, ofc7, prr, prr1, prr2, pvrl1, pvrl2, pvrr, pvrr1, pvrr2, sk-12
-
Chromosome Location:
Chr.7: 101305287 - 101336630 on Build UCB_Xtro_10.0
Chromosome Location:
Chr.Un|NW_016685664: 2774 - 6158 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
-
Assay ID Xt03683886_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: nectin2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001004860.1 NP_001004860.1 2 - 3 531 101
XM_012956717.2 XP_012812171.1 2 - 3 580 101
GenBank mRNA BC074691.1 - 2 - 3 531 101
CR762185.2 - 2 - 3 530 101
Gene Symbol: LOC108645444
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018090656.1 XP_017946145.1 1 - 2 350 101

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0