This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03684578_m1
See other ASF1B GE Assays ›
Gene Symbol:
asf1b
Gene Name:
anti-silencing function 1B histone chaperone
Gene Aliases:
asf1, cia-ii
Chromosome Location:
Chr.3: 123844292 - 123852227 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03684578_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: asf1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001004974.1 NP_001004974.1 2 - 3 207 99
GenBank mRNA BC075495.1 - 2 - 3 207 99
CR848421.2 - 2 - 3 243 99

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0