This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03684614_m1
See other PPIL3 GE Assays ›
Gene Symbol:
ppil3 LOC101733964
Gene Name:
peptidylprolyl isomerase like 3
peptidyl-prolyl cis-trans isomerase-like 3
Gene Aliases:
-
ppil3
Chromosome Location:
Chr.9: 3137899 - 3142639 on Build UCB_Xtro_10.0
Chromosome Location:
Chr.Un|NW_016685197: 7558 - 8981 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
-
Assay ID Xt03684614_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ppil3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001004983.1 NP_001004983.1 5 - 6 465 102
XM_012970297.2 XP_012825751.1 5 - 6 524 102
XM_012970298.2 XP_012825752.1 5 - 6 350 102
XM_018097081.1 XP_017952570.1 4 - 5 406 102
XM_018097082.1 XP_017952571.1 4 - 5 450 102
XM_018097083.1 XP_017952572.1 4 - 5 279 102
GenBank mRNA BC075537.1 - 5 - 6 465 102
Gene Symbol: LOC101733964
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA CR942529.1 - 4 - 5 302 102

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons

Panther Classification:

Molecular Function -

chaperone

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline