This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03684875_m1
See other UGT3A2 GE Assays ›
Gene Symbol:
ugt3a2 LOC100488176
Gene Name:
UDP glycosyltransferase 3 family, polypeptide A2
UDP-glucuronosyltransferase 3A1-like
Gene Aliases:
-
-
Chromosome Location:
Chr.1: 183711306 - 183722876 on Build UCB_Xtro_10.0
Chromosome Location:
Chr.1: 183697145 - 183710091 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
-
Assay ID Xt03684875_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ugt3a2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001005027.3 NP_001005027.3 5 - 6 971 75
XM_012959973.2 XP_012815427.1 5 - 6 1268 75
XM_018092265.1 XP_017947754.1 5 - 6 1235 75
XM_018092272.1 XP_017947761.1 5 - 6 1295 75
XM_018092275.1 XP_017947764.1 5 - 6 990 75
GenBank mRNA BC076709.1 - 5 - 6 960 75
CF218052.1 - 1 - 2 253 75
CU025035.1 - 5 - 6 940 75
Gene Symbol: LOC100488176
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004910430.3 XP_004910487.1 5 - 6 1517 75
XM_018090819.1 XP_017946308.1 5 - 6 979 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0