This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03684876_g1
See other UGT3A2 GE Assays ›
Gene Symbol:
ugt3a2
Gene Name:
UDP glycosyltransferase 3 family, polypeptide A2
Gene Aliases:
-
Chromosome Location:
Chr.1: 183711306 - 183722876 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03684876_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ugt3a2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001005027.3 NP_001005027.3 6 - 7 1206 86
XM_012959973.2 XP_012815427.1 6 - 7 1503 86
XM_018092265.1 XP_017947754.1 6 - 7 1470 86
XM_018092272.1 XP_017947761.1 6 - 7 1530 86
XM_018092275.1 XP_017947764.1 6 - 7 1225 86
GenBank mRNA BC076709.1 - 6 - 7 1195 86
BX687753.1 - 2 - 3 685 86
CF218052.1 - 2 - 3 488 86
CU025035.1 - 6 - 7 1175 86

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0