This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03685205_m1
See other TMEM183A GE Assays ›
Gene Symbol:
tmem183a
Gene Name:
transmembrane protein 183A
Gene Aliases:
-
Chromosome Location:
Chr.2: 68276459 - 68289803 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03685205_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: tmem183a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001005072.1 NP_001005072.1 5 - 6 789 100
XM_012960115.2 XP_012815569.1 5 - 6 729 100
XM_012960120.2 XP_012815574.1 4 - 5 883 100
XM_018091172.1 XP_017946661.1 5 - 6 866 100
GenBank mRNA BC076989.1 - 5 - 6 789 100
BC091580.1 - 4 - 5 613 100
CT030474.1 - 5 - 6 874 100

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0