This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03686039_s1
See other GJA1 GE Assays ›
Gene Symbol:
gja1
Gene Name:
gap junction protein alpha 1
Gene Aliases:
connexin43, cx43, dfnb38, gjal, oddd
Chromosome Location:
Chr.5: 33765417 - 33815818 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03686039_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: gja1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_203525.1 NP_988856.1 1 1051 106
XM_012962825.1 XP_012818279.1 2 1224 106
XM_018093818.1 XP_017949307.1 2 1175 106
GenBank mRNA AY043270.1 - 1 1051 106

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Xenopus tropicalis Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

gap junction

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline