This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03686335_m1
See other GPX3 GE Assays ›
Gene Symbol:
gpx3
Gene Name:
glutathione peroxidase 3
Gene Aliases:
gpx-p, gpx3-a, gpx3-b, gshpx-3, gshpx-p
Chromosome Location:
Chr.3: 5610545 - 5622646 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03686335_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: gpx3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_203630.2 NP_988961.2 1 - 2 204 101
GenBank mRNA BC061262.1 - 1 - 2 115 101
CR761198.2 - 1 - 2 95 101
CX855363.1 - 1 - 2 204 101

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC061262.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons

Panther Classification:

Molecular Function -

peroxidase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0