This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03686873_g1
See other LRTOMT GE Assays ›
Gene Symbol:
lrtomt
Gene Name:
leucine rich transmembrane and O-methyltransferase domain containing
Gene Aliases:
comt2, dfnb63, lrrc51, lrtomt1, lrtomt2
Chromosome Location:
Chr.Un|NW_016683486: 122562 - 126271 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03686873_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: lrtomt
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_203724.1 NP_989055.1 2 - 3 218 101
XM_012956643.2 XP_012812097.1 1 - 2 1027 101
XM_012956644.2 XP_012812098.1 2 - 3 979 101
XM_012956646.2 XP_012812100.1 2 - 3 447 101
XM_012956647.2 XP_012812101.1 2 - 3 364 101
XM_018089895.1 XP_017945384.1 2 - 3 123 101
GenBank mRNA BC061330.1 - 2 - 3 218 101

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline