This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03687304_g1
See other ENO1 GE Assays ›
Gene Symbol:
eno1
Gene Name:
enolase 1
Gene Aliases:
eno1-a, eno1-b, eno1l1, mbp-1, mpb1, nne, pph
Chromosome Location:
Chr.7: 80570063 - 80583742 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03687304_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: eno1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_203813.1 NP_989144.1 11 - 12 1269 71
XM_012966301.1 XP_012821755.1 11 - 12 1372 71
XM_012966302.1 XP_012821756.1 11 - 12 1346 71
XM_012966304.2 XP_012821758.1 11 - 12 1342 71
GenBank mRNA BC061287.1 - 11 - 12 1269 71
BC158432.1 - 11 - 12 1265 71
CT025514.2 - 11 - 12 1258 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons

Panther Classification:

Molecular Function -

lyase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline