Assay Details
| Gene Symbol: ldha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203822.1 | NP_989153.1 | 275 | 74 | ||
| GenBank mRNA | BC061615.1 | - | 275 | 74 | ||
| BC082741.1 | - | 281 | 74 | |||
| CT027954.1 | - | 265 | 74 | |||
More Information
Set Membership: |
|

ldha
Genomic Map