This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03688414_m1
See other EEF1G GE Assays ›
Gene Symbol:
eef1g
Gene Name:
eukaryotic translation elongation factor 1 gamma
Gene Aliases:
eef1g-a, eef1g-b, ef1g, gig35
Chromosome Location:
Chr.4: 41495683 - 41505044 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03688414_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: eef1g
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_203931.1 NP_989262.1 7 - 8 785 80
XM_012953583.1 XP_012809037.1 6 - 7 840 80
GenBank mRNA BC064264.1 - 7 - 8 785 80
BC080966.1 - 5 - 6 645 80
BC154938.1 - 5 - 6 583 80

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0