This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03690640_m1
See other ILF3 GE Assays ›
Gene Symbol:
ilf3
Gene Name:
interleukin enhancer binding factor 3
Gene Aliases:
4F.1, NF90, cbtf122, drbf, drbp76, ilf3-a, ilf3-b, mmp4, mphosph4, mpp4, nf-at-90, nfar, nfar-1, tcp80, ubp4, xilf3
Chromosome Location:
Chr.3: 127500456 - 127519015 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03690640_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ilf3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001005648.1 NP_001005648.1 5 - 6 746 70
XM_012953092.1 XP_012808546.1 5 - 6 791 70
XM_012953093.1 XP_012808547.1 5 - 6 820 70
XM_012953094.1 XP_012808548.1 4 - 5 619 70
XM_012953095.1 XP_012808549.1 5 - 6 791 70
XM_012953096.1 XP_012808550.1 5 - 6 791 70
XM_012953097.1 XP_012808551.1 5 - 6 791 70
XM_012953098.1 XP_012808552.1 5 - 6 791 70
GenBank mRNA BC074653.1 - 5 - 6 746 70
CT010548.1 - 4 - 5 616 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons

Panther Classification:

Molecular Function -

RNA metabolism protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline