This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03693510_m1
See other GABPB1 GE Assays ›
Gene Symbol:
gabpb1
Gene Name:
GA binding protein transcription factor subunit beta 1
Gene Aliases:
babpb2, e4tf1, e4tf1-47, e4tf1-53, e4tf1b, gabpb, gabpb1-a, gabpb1-b, gabpb2, nrf2b1, nrf2b2, xrpflbeta1
Chromosome Location:
Chr.3: 95006046 - 95025403 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03693510_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: gabpb1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001007483.1 NP_001007484.1 6 - 7 829 64
XM_012958861.2 XP_012814315.1 6 - 7 1028 64
XM_012958862.2 XP_012814316.1 6 - 7 1024 64
GenBank mRNA BC079923.1 - 6 - 7 829 64
CR942557.2 - 6 - 7 734 64

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline