This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03693952_m1
See other METTL9 GE Assays ›
Gene Symbol:
mettl9
Gene Name:
methyltransferase like 9
Gene Aliases:
-
Chromosome Location:
Chr.9: 19315129 - 19332335 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03693952_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: mettl9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001007898.1 NP_001007899.1 4 - 5 847 66
GenBank mRNA BC080334.1 - 4 - 5 847 66
CR760379.1 - 4 - 5 809 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline