This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03694637_g1
See other TADA3 GE Assays ›
Gene Symbol:
tada3
Gene Name:
transcriptional adaptor 3
Gene Aliases:
ada3, ngg1, tada3-a, tada3-b, tada3l
Chromosome Location:
Chr.4: 128008916 - 128018679 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03694637_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: tada3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001008039.1 NP_001008040.1 7 - 8 1155 98
XM_012960931.1 XP_012816385.1 7 - 8 1063 98
XM_018093018.1 XP_017948507.1 7 - 8 1032 98
GenBank mRNA BC080923.1 - 7 - 8 1155 98

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein histone modifying enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0