This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03698538_m1
See other MSI1 GE Assays ›
Gene Symbol:
msi1
Gene Name:
musashi RNA binding protein 1
Gene Aliases:
Msi1h, Musashi, Musashi-1, Musashi1, XNrp-1, nrp-1B
Chromosome Location:
Chr.1: 143109567 - 143133605 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03698538_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: msi1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001011470.1 NP_001011470.1 14 - 15 1053 65
XM_012961432.2 XP_012816886.1 14 - 15 1123 65
XM_012961437.2 XP_012816891.1 13 - 14 1067 65
XM_012961440.2 XP_012816894.1 13 - 14 1046 65
GenBank mRNA BC088603.1 - 14 - 15 1053 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0