Assay Details
| Gene Symbol: polr2k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016509.1 | NP_001016509.1 | 2 - 3 | 271 | 111 | |
| XM_012964704.2 | XP_012820158.1 | 3 - 4 | 309 | 111 | ||
| XM_012964705.2 | XP_012820159.1 | 3 - 4 | 329 | 111 | ||
| XM_018094653.1 | XP_017950142.1 | 3 - 4 | 287 | 111 | ||
| GenBank mRNA | BC170861.1 | - | 2 - 3 | 201 | 111 | |
| BC170863.1 | - | 2 - 3 | 201 | 111 | ||
| CR761743.2 | - | 3 - 4 | 243 | 111 | ||
| CR926237.1 | - | 2 - 3 | 271 | 111 | ||
More Information
Set Membership: |
|

polr2k
Genomic Map