This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03705702_m1
See other AKR1A1 GE Assays ›
Gene Symbol:
akr1a1
Gene Name:
aldo-keto reductase family 1, member A1 (aldehyde reductase)
Gene Aliases:
aldr1, alr, arm, dd3
Chromosome Location:
Chr.4: 73275234 - 73297557 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03705702_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: akr1a1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001016137.2 NP_001016137.1 8 - 9 1150 81
XM_012961063.2 XP_012816517.1 7 - 8 909 81
XM_012961064.2 XP_012816518.1 7 - 8 885 81
XM_012961066.2 XP_012816520.1 8 - 9 1142 81
XM_012961067.2 XP_012816521.1 8 - 9 1034 81
XM_018093036.1 XP_017948525.1 8 - 9 1089 81
XM_018093037.1 XP_017948526.1 7 - 8 1278 81
GenBank mRNA BC170750.1 - 7 - 8 877 81
BC170752.1 - 7 - 8 877 81
CR762028.2 - 8 - 9 1150 81

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons

Panther Classification:

Molecular Function -

oxidoreductase reductase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0