This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03706208_m1
See other PPP1R14B GE Assays ›
Gene Symbol:
ppp1r14b
Gene Name:
protein phosphatase 1 regulatory inhibitor subunit 14B
Gene Aliases:
phi-1, plcb3n, png, som172
Chromosome Location:
Chr.4: 40198556 - 40204921 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03706208_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ppp1r14b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001016307.2 NP_001016307.1 3 - 4 327 107
GenBank mRNA CR761497.2 - 3 - 4 327 107

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Xenopus tropicalis Probe spans exons

Panther Classification:

Molecular Function -

phosphatase modulator

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-fxmhk:80/100.66.79.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0