This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03706521_m1
See other UBE2I GE Assays ›
Gene Symbol:
ube2i
Gene Name:
ubiquitin conjugating enzyme E2 I
Gene Aliases:
c358b7.1, p18, ubc9, ubce9
Chromosome Location:
Chr.9: 22910955 - 22925672 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03706521_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ube2i
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001016408.2 NP_001016408.1 5 - 6 418 76
XM_012970417.2 XP_012825871.1 6 - 7 751 76
XM_012970418.2 XP_012825872.1 6 - 7 797 76
XM_012970419.2 XP_012825873.1 7 - 8 811 76
GenBank mRNA CR761123.2 - 5 - 6 418 76
CR926257.1 - 6 - 7 494 76

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Xenopus tropicalis Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kt9qp:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0