This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03708123_m1
See other PTPN11 GE Assays ›
Gene Symbol:
ptpn11
Gene Name:
protein tyrosine phosphatase, non-receptor type 11
Gene Aliases:
-
Chromosome Location:
Chr.1: 155206854 - 155235769 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03708123_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ptpn11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001016832.2 NP_001016832.1 2 - 3 314 69
XM_012962883.2 XP_012818337.1 2 - 3 366 69
XM_018093512.1 XP_017949001.1 3 - 4 713 69
GenBank mRNA CR855502.2 - 2 - 3 314 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Xenopus tropicalis Probe spans exons

Panther Classification:

Molecular Function -

protein phosphatase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4krp5:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0