Assay Details
| Gene Symbol: washc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016854.2 | NP_001016854.1 | 6 - 7 | 949 | 64 | |
| GenBank mRNA | BC123023.1 | - | 6 - 7 | 917 | 64 | |
| CR855419.2 | - | 6 - 7 | 949 | 64 | ||
More Information
Set Membership: |
|

washc1
Genomic Map