Assay Details
| Gene Symbol: zmpste24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017255.2 | NP_001017255.1 | 6 - 7 | 761 | 74 | |
| XM_012957049.2 | XP_012812503.1 | 6 - 7 | 1283 | 74 | ||
| GenBank mRNA | CR760049.2 | - | 6 - 7 | 761 | 74 | |
More Information
Set Membership: |
|

zmpste24
Genomic Map