This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03710008_m1
See other ELAVL3 GE Assays ›
Gene Symbol:
elavl3
Gene Name:
ELAV like neuron-specific RNA binding protein 3
Gene Aliases:
elrC, huc, hucl, ple21
Chromosome Location:
Chr.3: 128354707 - 128385960 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03710008_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: elavl3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001030326.2 NP_001025497.1 2 - 3 377 85
XM_012953160.2 XP_012808614.1 2 - 3 712 85
XM_012953161.2 XP_012808615.1 2 - 3 712 85
XM_012953162.2 XP_012808616.1 2 - 3 709 85
XM_012953163.2 XP_012808617.1 2 - 3 712 85
XM_012953164.2 XP_012808618.1 2 - 3 711 85
XM_012953165.2 XP_012808619.1 2 - 3 711 85
XM_012953166.2 XP_012808620.1 2 - 3 708 85
XM_012953167.2 XP_012808621.1 2 - 3 710 85
XM_012953168.2 XP_012808622.1 2 - 3 708 85
XM_018092117.1 XP_017947606.1 2 - 3 711 85
GenBank mRNA BC080177.1 - 2 - 3 698 85
BC136134.1 - 2 - 3 683 85
CR761704.2 - 2 - 3 377 85

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons

Panther Classification:

Molecular Function -

translation initiation factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0