This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03710310_m1
See other RPS6KB1 GE Assays ›
Gene Symbol:
rps6kb1
Gene Name:
ribosomal protein S6 kinase B1
Gene Aliases:
p70-alpha, p70-s6k, p70s6k, ps6k, rps6ka1, rps6kb1-A, s6K1, s6k, stk14a
Chromosome Location:
Chr.2: 87405657 - 87435204 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03710310_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: rps6kb1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001039728.1 NP_001034817.1 10 - 11 1010 66
XM_012956674.2 XP_012812128.1 10 - 11 1026 66
XM_018091149.1 XP_017946638.1 10 - 11 1026 66
GenBank mRNA BC064239.1 - 10 - 11 984 66
CR848655.2 - 10 - 11 1010 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons

Panther Classification:

Molecular Function -

protein modifying enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline