This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03710812_m1
See other NAXD GE Assays ›
Gene Symbol:
naxd
Gene Name:
NAD(P)HX dehydratase
Gene Aliases:
carkd
Chromosome Location:
Chr.2: 113588482 - 113605453 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03710812_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: naxd
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001016310.4 NP_001016310.2 10 - 11 1878 73
NM_001247985.1 NP_001234914.1 9 - 10 890 73
XM_012956947.2 XP_012812401.1 9 - 10 991 73
XM_012956948.2 XP_012812402.1 9 - 10 1081 73
XM_018091246.1 XP_017946735.1 9 - 10 3723 73
XM_018091247.1 XP_017946736.1 9 - 10 3461 73
GenBank mRNA BC121541.1 - 10 - 11 1838 73
CR761491.2 - 10 - 11 1306 73
DT410042.1 - 2 - 3 188 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons

Panther Classification:

Molecular Function -

dehydratase lyase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0