This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03711010_m1
See other ELAVL2 GE Assays ›
Gene Symbol:
elavl2
Gene Name:
ELAV like neuron-specific RNA binding protein 2
Gene Aliases:
el-1, elavl2-a, elavl2-b, elrB, hel-n1, heln1, hub
Chromosome Location:
Chr.1: 109533069 - 109623625 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03711010_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: elavl2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001030327.4 NP_001025498.3 5 - 6 738 93
XM_012963568.2 XP_012819022.1 6 - 7 721 93
XM_012963570.1 XP_012819024.1 6 - 7 1784 93
XM_012963574.2 XP_012819028.1 5 - 6 746 93
XM_012963578.1 XP_012819032.1 5 - 6 1781 93
XM_012963582.2 XP_012819036.1 6 - 7 721 93
XM_012963585.2 XP_012819039.1 5 - 6 733 93
XM_012963590.2 XP_012819044.1 5 - 6 630 93
XM_012963597.2 XP_012819051.1 4 - 5 642 93
XM_012963602.2 XP_012819056.1 6 - 7 721 93
XM_012963604.2 XP_012819058.1 5 - 6 782 93
XM_018093799.1 XP_017949288.1 6 - 7 712 93
XM_018093803.1 XP_017949292.1 6 - 7 718 93
XM_018093807.1 XP_017949296.1 6 - 7 787 93
GenBank mRNA BC127338.1 - 5 - 6 707 93
CR761432.2 - 4 - 5 588 93

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons

Panther Classification:

Molecular Function -

translation initiation factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nbrrd:80/100.66.77.69:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0