This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03712219_m1
See other NUDT22 GE Assays ›
Gene Symbol:
nudt22
Gene Name:
nudix hydrolase 22
Gene Aliases:
-
Chromosome Location:
Chr.4: 40241049 - 40260631 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03712219_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: nudt22
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001045632.1 NP_001039097.1 3 - 4 709 71
XM_012961262.2 XP_012816716.1 3 - 4 776 71
XM_012961264.1 XP_012816718.1 3 - 4 1040 71
GenBank mRNA CR942377.2 - 3 - 4 709 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Xenopus tropicalis Probe spans exons

Panther Classification:

Molecular Function -

phosphatase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0