This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03712228_g1
See other ZNF160 GE Assays ›
Gene Symbol:
znf160
Gene Name:
zinc finger protein 160
Gene Aliases:
f11, hzf5, kr18
Chromosome Location:
Chr.8: 54782517 - 54792412 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03712228_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: znf160
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001045635.1 NP_001039100.1 1 - 2 200 94
XM_012968546.2 XP_012824000.1 2 - 3 274 94
XM_012968547.2 XP_012824001.1 2 - 3 308 94
XM_012968548.2 XP_012824002.1 2 - 3 297 94
XM_012968549.2 XP_012824003.1 2 - 3 348 94
XM_018096289.1 XP_017951778.1 2 - 3 274 94
GenBank mRNA BC154853.1 - 1 - 2 183 94
CR942385.2 - 1 - 2 200 94

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0