This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03713205_m1
See other OGFOD2 GE Assays ›
Gene Symbol:
ogfod2
Gene Name:
2-oxoglutarate and iron dependent oxygenase domain containing 2
Gene Aliases:
-
Chromosome Location:
Chr.1: 152745683 - 152756725 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03713205_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ogfod2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001045746.1 NP_001039211.1 6 - 7 794 100
XM_012964506.2 XP_012819960.1 6 - 7 1152 100
XM_012964510.2 XP_012819964.1 5 - 6 693 100
XM_012964515.2 XP_012819969.1 7 - 8 986 100
XM_012964518.2 XP_012819972.1 4 - 5 579 100
XM_018094113.1 XP_017949602.1 6 - 7 759 100
XM_018094117.1 XP_017949606.1 6 - 7 681 100
XM_018094120.1 XP_017949609.1 5 - 6 648 100
GenBank mRNA BC157323.1 - 7 - 8 991 100
CR942500.2 - 6 - 7 794 100

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons

Panther Classification:

Molecular Function -

RNA metabolism protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0