Assay Details
| Gene Symbol: ppp2r3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079206.1 | NP_001072674.1 | 5 - 6 | 776 | 103 | |
| XM_012957298.2 | XP_012812752.1 | 4 - 5 | 1003 | 103 | ||
| GenBank mRNA | BC124001.1 | - | 5 - 6 | 776 | 103 | |
More Information
Set Membership: |
|

ppp2r3b
Genomic Map