This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03718050_m1
See other KMT5A GE Assays ›
Gene Symbol:
kmt5a
Gene Name:
lysine methyltransferase 5A
Gene Aliases:
pr-set7, set07, set8, setd8
Chromosome Location:
Chr.1: 152441877 - 152454784 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03718050_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: kmt5a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001079347.1 NP_001072815.1 7 - 8 943 98
XM_018094658.1 XP_017950147.1 6 - 7 891 98
GenBank mRNA BC121601.1 - 7 - 8 943 98

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons

Panther Classification:

Molecular Function -

protein modifying enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0