This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03719101_m1
See other FGFR1 GE Assays ›
Gene Symbol:
fgfr1
Gene Name:
fibroblast growth factor receptor 1
Gene Aliases:
X1FGFR, XFGFR-1, bfgfr, cd331, cek, fgfbr, fgfr-1, fgfr1-a, fgfr1-b, flg, flt-2, flt2, hbgfr, kal2, n-sam, ogd, xfgfr1
Chromosome Location:
Chr.3: 120588146 - 120635606 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03719101_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: fgfr1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001015894.2 NP_001015894.1 6 - 7 1146 66
XM_012958974.2 XP_012814428.1 6 - 7 1890 66
XM_012958975.2 XP_012814429.1 6 - 7 1890 66
XM_012958976.2 XP_012814430.1 6 - 7 1890 66
XM_012958977.2 XP_012814431.1 5 - 6 1626 66
XM_012958978.2 XP_012814432.1 5 - 6 1626 66
GenBank mRNA BC135670.1 - 6 - 7 1146 66
CR759996.2 - 6 - 7 867 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons

Panther Classification:

Molecular Function -

transmembrane signal receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-f8k5r:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline