Assay Details
| Gene Symbol: znf180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079065.1 | NP_001072533.1 | 3 - 4 | 309 | 89 | |
| XM_012959247.1 | XP_012814701.1 | 3 - 4 | 492 | 89 | ||
| XM_012959248.1 | XP_012814702.1 | 3 - 4 | 476 | 89 | ||
| XM_012959249.1 | XP_012814703.1 | 3 - 4 | 392 | 89 | ||
| XM_012959250.2 | XP_012814704.1 | 3 - 4 | 310 | 89 | ||
| XM_018092172.1 | XP_017947661.1 | 3 - 4 | 323 | 89 | ||
| GenBank mRNA | BC122022.1 | - | 3 - 4 | 309 | 89 | |
| CU075330.1 | - | 3 - 4 | 339 | 89 | ||
More Information
Set Membership: |
|

znf180
Genomic Map